Assay Details
| Gene Symbol: VHA-A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127713.4 | NP_179736.1 | 17 - 18 | 2449 | 82 | |
| GenBank mRNA | AF424556.1 | - | 17 - 18 | 2298 | 82 | |
| AY099690.1 | - | 17 - 18 | 2341 | 82 | ||
More Information
Set Membership: |
|

VHA-A2
Genomic Map