Assay Details
| Gene Symbol: VHA-A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127713.4 | NP_179736.1 | 7 - 8 | 1042 | 72 | |
| GenBank mRNA | AF424556.1 | - | 7 - 8 | 891 | 72 | |
| AY099690.1 | - | 7 - 8 | 934 | 72 | ||
More Information
Set Membership: |
|

VHA-A2
Genomic Map