Assay Details
| Gene Symbol: SAE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202641.1 | NP_001189570.1 | 5 - 6 | 716 | 83 | |
| NM_127719.4 | NP_179742.2 | 5 - 6 | 700 | 83 | ||
| NM_201777.1 | NP_973506.1 | 5 - 6 | 691 | 83 | ||
| GenBank mRNA | AK117731.1 | - | 5 - 6 | 691 | 83 | |
| BT044606.1 | - | 5 - 6 | 533 | 83 | ||
More Information
Set Membership: |
|

SAE2
Genomic Map