Assay Details
| Gene Symbol: SAE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202641.1 | NP_001189570.1 | 7 - 8 | 1410 | 131 | |
| NM_127719.4 | NP_179742.2 | 7 - 8 | 1385 | 131 | ||
| NM_201777.1 | NP_973506.1 | 7 - 8 | 1376 | 131 | ||
| GenBank mRNA | AK117731.1 | - | 7 - 8 | 1376 | 131 | |
| BT044606.1 | - | 7 - 8 | 1218 | 131 | ||
More Information
Set Membership: |
|

SAE2
Genomic Map