Assay Details
| Gene Symbol: GRP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127738.5 | NP_179760.1 | 1 - 2 | 349 | 74 | |
| NM_179686.2 | NP_850017.1 | 1 - 2 | 185 | 74 | ||
| GenBank mRNA | AF428381.1 | - | 1 - 2 | 165 | 74 | |
| AY054284.1 | - | 1 - 2 | 165 | 74 | ||
| L00648.1 | - | 1 - 2 | 148 | 74 | ||
| Z14987.1 | - | 148 | 74 | |||
| Gene Symbol: AT3G13230 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK229503.1 | - | 165 | 74 | ||
More Information
Set Membership: |
|

GRP7
Genomic Map