Assay Details
| Gene Symbol: AAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036317.1 | NP_001031394.1 | 2 - 3 | 230 | 84 | |
| NM_127791.3 | NP_565529.1 | 1 - 2 | 733 | 84 | ||
| NM_179691.4 | NP_850022.1 | 2 - 3 | 166 | 84 | ||
| GenBank mRNA | AY064152.1 | - | 1 - 2 | 215 | 84 | |
| AY124811.1 | - | 1 - 2 | 122 | 84 | ||
| HM638413.1 | - | 1 - 2 | 122 | 84 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_179691.4 transcripts.
Set Membership: |
|

AAT
Genomic Map