Assay Details
| Gene Symbol: ALKBH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001335786.1 | NP_001325000.1 | 5 - 6 | 974 | 86 | |
| NM_001335787.1 | NP_001324995.1 | 6 - 7 | 819 | 86 | ||
| NM_001335788.1 | NP_001324999.1 | 6 - 7 | 864 | 86 | ||
| NM_001335789.1 | NP_001324998.1 | 6 - 7 | 819 | 86 | ||
| NM_001335790.1 | NP_001324997.1 | 6 - 7 | 779 | 86 | ||
| NM_001335791.1 | NP_001324996.1 | 5 - 6 | 810 | 86 | ||
| NM_001335792.1 | NP_001324994.1 | 6 - 7 | 700 | 86 | ||
| NM_001335793.1 | NP_001324993.1 | 3 - 4 | 516 | 86 | ||
| NM_127792.4 | NP_565530.1 | 6 - 7 | 819 | 86 | ||
More Information
Set Membership: |
|

ALKBH2
Genomic Map