Assay Details
Gene Symbol: AT2G22370 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127802.4 | NP_565534.1 | 3 - 4 | 329 | 89 | |
GenBank mRNA | BT029378.1 | - | 3 - 4 | 197 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
