Assay Details
Gene Symbol: AT2G22370 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127802.4 | NP_565534.1 | 4 - 5 | 687 | 69 | |
GenBank mRNA | BT029378.1 | - | 4 - 5 | 555 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
