Assay Details
Gene Symbol: GEM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127813.3 | NP_565538.1 | 2 - 3 | 838 | 82 | |
NM_201781.1 | NP_973510.1 | 2 - 3 | 565 | 82 | ||
GenBank mRNA | EF490993.1 | - | 2 - 3 | 443 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
