Assay Details
Gene Symbol: CYCD2;1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202647.1 | NP_001189576.1 | 1 - 2 | 586 | 73 | |
NM_127815.4 | NP_179835.1 | 1 - 2 | 608 | 73 | ||
GenBank mRNA | BT025242.1 | - | 1 - 2 | 314 | 73 | |
X83370.1 | - | 508 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
