Assay Details
| Gene Symbol: NPY4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001335835.1 | NP_001325415.1 | 3 - 4 | 1175 | 70 | |
| NM_127869.3 | NP_179887.1 | 3 - 4 | 1343 | 70 | ||
| GenBank mRNA | AK117486.1 | - | 3 - 4 | 1297 | 70 | |
| AY102541.1 | - | 3 - 4 | 1328 | 70 | ||
| AY954795.1 | - | 3 - 4 | 1200 | 70 | ||
| BT005613.1 | - | 3 - 4 | 1200 | 70 | ||
More Information
Set Membership: |
|

NPY4
Genomic Map