Assay Details
Gene Symbol: PAB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127899.4 | NP_179916.1 | 2 - 3 | 705 | 149 | |
GenBank mRNA | AY050859.1 | - | 2 - 3 | 565 | 149 | |
AY079389.1 | - | 2 - 3 | 411 | 149 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
