Assay Details
Gene Symbol: XLG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127910.3 | NP_565553.1 | 1 - 2 | 1312 | 153 | |
GenBank mRNA | AF060941.1 | - | 1 - 2 | 1236 | 153 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
