Assay Details
| Gene Symbol: AT2G23700 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127933.3 | NP_179950.1 | 10 - 11 | 2296 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT2G23700| Gene Symbol: AT2G23700 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127933.3 | NP_179950.1 | 10 - 11 | 2296 | 65 | |
Set Membership: |
|