Assay Details
Gene Symbol: SCA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127973.5 | NP_179989.1 | 10 - 11 | 2246 | 146 | |
NM_201796.2 | NP_973525.2 | 10 - 11 | 2246 | 146 | ||
GenBank mRNA | AK229652.1 | - | 10 - 11 | 2152 | 146 | |
Y08463.1 | - | 2193 | 146 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
