Assay Details
Gene Symbol: SCA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127973.5 | NP_179989.1 | 12 - 13 | 2392 | 62 | |
NM_201796.2 | NP_973525.2 | 12 - 13 | 2392 | 62 | ||
GenBank mRNA | AK229652.1 | - | 12 - 13 | 2298 | 62 | |
Y08463.1 | - | 2339 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
