Assay Details
| Gene Symbol: AT2G24340 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127996.1 | NP_180011.1 | 4 - 5 | 754 | 116 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT2G24340| Gene Symbol: AT2G24340 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127996.1 | NP_180011.1 | 4 - 5 | 754 | 116 | |
Set Membership: |
|