Assay Details
Gene Symbol: WSIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202976.1 | NP_001189905.1 | 10 - 11 | 1431 | 90 | |
NM_001338190.1 | NP_001325883.1 | 9 - 10 | 1663 | 90 | ||
NM_112458.5 | NP_188209.3 | 10 - 11 | 1427 | 90 | ||
NM_180672.4 | NP_851003.2 | 10 - 11 | 1431 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
