Assay Details
| Gene Symbol: OASC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_180691.1 | NP_851022.1 | 10 - 11 | 1355 | 73 | |
| GenBank mRNA | AK316875.1 | - | 10 - 11 | 1336 | 73 | |
More Information
Set Membership: |
|

OASC
Genomic Map