Assay Details
| Gene Symbol: OASC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115838.6 | NP_191535.2 | 9 - 10 | 1308 | 67 | |
| NM_180691.1 | NP_851022.1 | 9 - 10 | 1271 | 67 | ||
| GenBank mRNA | AK226285.1 | - | 9 - 10 | 1256 | 67 | |
| AK316875.1 | - | 9 - 10 | 1252 | 67 | ||
| AY099721.1 | - | 9 - 10 | 1270 | 67 | ||
| AY128885.1 | - | 9 - 10 | 1200 | 67 | ||
More Information
Set Membership: |
|

OASC
Genomic Map