Assay Details
| Gene Symbol: OASC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_180692.1 | NP_851023.1 | 9 - 10 | 1284 | 99 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

OASC| Gene Symbol: OASC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_180692.1 | NP_851023.1 | 9 - 10 | 1284 | 99 | |
Set Membership: |
|