Assay Details
| Gene Symbol: HD2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_122171.3 | NP_197657.3 | 2 - 3 | 186 | 65 | |
| NM_180725.3 | NP_851056.1 | 2 - 3 | 257 | 65 | ||
| GenBank mRNA | AF044914.1 | - | 1 - 2 | 83 | 65 | |
| AF195546.1 | - | 2 - 3 | 143 | 65 | ||
| AY059893.1 | - | 2 - 3 | 157 | 65 | ||
| AY079014.1 | - | 2 - 3 | 155 | 65 | ||
| AY122890.1 | - | 1 - 2 | 83 | 65 | ||
More Information
Set Membership: |
|

HD2B
Genomic Map