Assay Details
| Gene Symbol: PHS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036856.2 | NP_001031933.1 | 9 - 10 | 2563 | 78 | |
| NM_122277.2 | NP_197761.2 | 10 - 11 | 2456 | 78 | ||
| NM_180735.4 | NP_851066.2 | 10 - 11 | 2545 | 78 | ||
| GenBank mRNA | AB161693.1 | - | 10 - 11 | 2512 | 78 | |
More Information
Set Membership: |
|

PHS1
Genomic Map