Assay Details
| Gene Symbol: TGG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036863.1 | NP_001031940.1 | 10 - 11 | 1298 | 98 | |
| NM_122499.4 | NP_568479.1 | 10 - 11 | 1317 | 98 | ||
| NM_180745.2 | NP_851076.2 | 10 - 11 | 1298 | 98 | ||
| GenBank mRNA | AF360348.1 | - | 10 - 11 | 1295 | 98 | |
| AY113880.1 | - | 10 - 11 | 1295 | 98 | ||
More Information
Set Membership: |
|

TGG2
Genomic Map