Assay Details
| Gene Symbol: TGG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_122501.3 | NP_197972.2 | 7 - 8 | 949 | 82 | |
| NM_180746.3 | NP_851077.1 | 7 - 8 | 949 | 82 | ||
| GenBank mRNA | AY045681.1 | - | 7 - 8 | 943 | 82 | |
| AY090382.1 | - | 7 - 8 | 912 | 82 | ||
| L11454.1 | - | 7 - 8 | 945 | 82 | ||
More Information
Set Membership: |
|

TGG1
Genomic Map