Assay Details
| Gene Symbol: HDA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_122505.4 | NP_568480.2 | 9 - 10 | 986 | 91 | |
| NM_180747.2 | NP_851078.1 | 9 - 10 | 856 | 91 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HDA2| Gene Symbol: HDA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_122505.4 | NP_568480.2 | 9 - 10 | 986 | 91 | |
| NM_180747.2 | NP_851078.1 | 9 - 10 | 856 | 91 | ||
Set Membership: |
|