Assay Details
Gene Symbol: MK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203482.1 | NP_001190411.1 | 3 - 4 | 453 | 85 | |
NM_122627.6 | NP_198097.1 | 2 - 3 | 1004 | 85 | ||
NM_180753.4 | NP_851084.1 | 3 - 4 | 471 | 85 | ||
GenBank mRNA | AY093165.1 | - | 3 - 4 | 428 | 85 | |
BT002104.1 | - | 2 - 3 | 381 | 85 | ||
X77793.1 | - | 703 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
