Assay Details
| Gene Symbol: SPL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123693.6 | NP_199141.1 | 3 - 4 | 999 | 120 | |
| NM_180791.3 | NP_851122.1 | 3 - 4 | 1634 | 120 | ||
| NM_203146.4 | NP_974875.1 | 3 - 4 | 1550 | 120 | ||
| GenBank mRNA | AJ011626.1 | - | 3 - 4 | 806 | 120 | |
| AK230160.1 | - | 3 - 4 | 892 | 120 | ||
| BT025984.1 | - | 2 - 3 | 753 | 120 | ||
More Information
Set Membership: |
|

SPL2
Genomic Map