Assay Details
| Gene Symbol: SPL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123693.6 | NP_199141.1 | 4 - 5 | 1213 | 64 | |
| NM_180791.3 | NP_851122.1 | 4 - 5 | 1848 | 64 | ||
| NM_203146.4 | NP_974875.1 | 4 - 5 | 1764 | 64 | ||
| GenBank mRNA | AJ011626.1 | - | 4 - 5 | 1020 | 64 | |
| AK230160.1 | - | 4 - 5 | 1106 | 64 | ||
| BT025984.1 | - | 3 - 4 | 967 | 64 | ||
More Information
Set Membership: |
|

SPL2
Genomic Map