Assay Details
Gene Symbol: FRA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124156.5 | NP_199593.2 | 20 - 21 | 2945 | 141 | |
NM_180820.2 | NP_851151.1 | 20 - 21 | 2966 | 141 | ||
GenBank mRNA | AY158083.1 | - | 20 - 21 | 2616 | 141 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
