Assay Details
| Gene Symbol: FRA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124156.5 | NP_199593.2 | 9 - 10 | 1713 | 78 | |
| NM_180820.2 | NP_851151.1 | 9 - 10 | 1734 | 78 | ||
| GenBank mRNA | AY158083.1 | - | 9 - 10 | 1384 | 78 | |
More Information
Set Membership: |
|

FRA1
Genomic Map