Assay Details
Gene Symbol: ACA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345242.1 | NP_001331281.1 | 16 - 17 | 1578 | 70 | |
NM_125093.6 | NP_200521.3 | 16 - 17 | 1627 | 70 | ||
NM_180869.3 | NP_851200.1 | 16 - 17 | 1469 | 70 | ||
GenBank mRNA | AJ249352.1 | - | 16 - 17 | 1471 | 70 | |
AK226961.1 | - | 15 - 16 | 1393 | 70 | ||
AY069869.1 | - | 16 - 17 | 1471 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
