Assay Details
Gene Symbol: ACA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345242.1 | NP_001331281.1 | 17 - 18 | 1618 | 75 | |
NM_125093.6 | NP_200521.3 | 17 - 18 | 1667 | 75 | ||
NM_180869.3 | NP_851200.1 | 17 - 18 | 1509 | 75 | ||
GenBank mRNA | AJ249352.1 | - | 17 - 18 | 1511 | 75 | |
AK226961.1 | - | 16 - 17 | 1433 | 75 | ||
AY069869.1 | - | 17 - 18 | 1511 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
