Assay Details
| Gene Symbol: RHD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115005.5 | NP_190714.2 | 13 - 14 | 1401 | 108 | |
| GenBank mRNA | AF266459.1 | - | 12 - 13 | 1027 | 108 | |
| AF360267.1 | - | 13 - 14 | 1121 | 108 | ||
| AY040078.1 | - | 12 - 13 | 1027 | 108 | ||
| AY227250.1 | - | 12 - 13 | 1027 | 108 | ||
More Information
Set Membership: |
|

RHD4
Genomic Map