Assay Details
| Gene Symbol: RHD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115005.5 | NP_190714.2 | 16 - 17 | 1707 | 96 | |
| GenBank mRNA | AF266459.1 | - | 15 - 16 | 1333 | 96 | |
| AF360267.1 | - | 16 - 17 | 1427 | 96 | ||
| AY040078.1 | - | 15 - 16 | 1333 | 96 | ||
| AY227250.1 | - | 15 - 16 | 1333 | 96 | ||
More Information
Set Membership: |
|

RHD4
Genomic Map