Assay Details
| Gene Symbol: RHD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115005.5 | NP_190714.2 | 3 - 4 | 604 | 65 | |
| GenBank mRNA | AF266459.1 | - | 2 - 3 | 230 | 65 | |
| AF360267.1 | - | 3 - 4 | 324 | 65 | ||
| AY040078.1 | - | 2 - 3 | 230 | 65 | ||
| AY227250.1 | - | 2 - 3 | 230 | 65 | ||
More Information
Set Membership: |
|

RHD4
Genomic Map