Assay Details
| Gene Symbol: GTE6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203143.1 | NP_001190072.1 | 4 - 5 | 537 | 95 | |
| NM_115088.2 | NP_190796.1 | 3 - 4 | 527 | 95 | ||
More Information
Set Membership: |
|

GTE6
Genomic Map