Assay Details
| Gene Symbol: PAD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115103.4 | NP_190811.1 | 1 - 2 | 765 | 89 | |
| GenBank mRNA | AF188329.1 | - | 1 - 2 | 294 | 89 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PAD4| Gene Symbol: PAD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115103.4 | NP_190811.1 | 1 - 2 | 765 | 89 | |
| GenBank mRNA | AF188329.1 | - | 1 - 2 | 294 | 89 | |
Set Membership: |
|