Assay Details
Gene Symbol: BGAL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339590.1 | NP_001325739.1 | 10 - 11 | 1286 | 86 | |
NM_115144.4 | NP_190852.2 | 10 - 11 | 1248 | 86 | ||
GenBank mRNA | AF367327.1 | - | 10 - 11 | 1180 | 86 | |
AJ270298.1 | - | 10 - 11 | 1137 | 86 | ||
BT000511.1 | - | 10 - 11 | 1091 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
