Assay Details
Gene Symbol: BGAL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339590.1 | NP_001325739.1 | 17 - 18 | 2271 | 105 | |
NM_115144.4 | NP_190852.2 | 17 - 18 | 2233 | 105 | ||
GenBank mRNA | AF367327.1 | - | 17 - 18 | 2165 | 105 | |
AJ270298.1 | - | 17 - 18 | 2122 | 105 | ||
BT000511.1 | - | 17 - 18 | 2076 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
