Assay Details
| Gene Symbol: PAL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115186.4 | NP_190894.1 | 1 - 2 | 897 | 111 | |
| GenBank mRNA | AF367308.1 | - | 1 - 2 | 506 | 111 | |
| AY133595.1 | - | 1 - 2 | 402 | 111 | ||
| AY303129.1 | - | 1 - 2 | 402 | 111 | ||
More Information
Set Membership: |
|

PAL2
Genomic Map