Assay Details
| Gene Symbol: PUMP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115271.5 | NP_190979.1 | 3 - 4 | 591 | 82 | |
| GenBank mRNA | AF146226.1 | - | 3 - 4 | 371 | 82 | |
| AJ001264.1 | - | 3 - 4 | 533 | 82 | ||
| AY056272.1 | - | 3 - 4 | 552 | 82 | ||
| AY091185.1 | - | 3 - 4 | 371 | 82 | ||
More Information
Set Membership: |
|

PUMP1
Genomic Map