Assay Details
| Gene Symbol: PUMP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115271.5 | NP_190979.1 | 4 - 5 | 723 | 91 | |
| GenBank mRNA | AF146226.1 | - | 4 - 5 | 503 | 91 | |
| AY056272.1 | - | 4 - 5 | 684 | 91 | ||
| AY091185.1 | - | 4 - 5 | 503 | 91 | ||
More Information
Set Membership: |
|

PUMP1
Genomic Map