Assay Details
| Gene Symbol: PUMP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115271.5 | NP_190979.1 | 5 - 6 | 827 | 68 | |
| GenBank mRNA | AF146226.1 | - | 5 - 6 | 607 | 68 | |
| AY056272.1 | - | 5 - 6 | 788 | 68 | ||
| AY091185.1 | - | 5 - 6 | 607 | 68 | ||
More Information
Set Membership: |
|

PUMP1
Genomic Map