Assay Details
Gene Symbol: CDKB1;1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115278.3 | NP_190986.1 | 3 - 4 | 1047 | 74 | |
GenBank mRNA | AY084810.1 | - | 3 - 4 | 923 | 74 | |
BT026079.1 | - | 3 - 4 | 836 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
