Assay Details
| Gene Symbol: RGD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203156.1 | NP_001190085.1 | 13 - 14 | 2181 | 117 | |
| NM_115288.8 | NP_190996.3 | 12 - 13 | 2212 | 117 | ||
| GenBank mRNA | AB440793.1 | - | 11 - 12 | 1950 | 117 | |
More Information
Set Membership: |
|

RGD3
Genomic Map