Assay Details
Gene Symbol: SKIP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339669.1 | NP_001326222.1 | 5 - 6 | 575 | 70 | |
NM_001339670.1 | NP_001326221.1 | 3 - 4 | 476 | 70 | ||
NM_115304.3 | NP_566999.1 | 4 - 5 | 638 | 70 | ||
GenBank mRNA | AK227454.1 | - | 4 - 5 | 464 | 70 | |
BT005863.1 | - | 4 - 5 | 446 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
