Assay Details
| Gene Symbol: HAG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115318.3 | NP_567002.1 | 10 - 11 | 1596 | 99 | |
| GenBank mRNA | AF338768.1 | - | 10 - 11 | 1426 | 99 | |
| BT002057.1 | - | 10 - 11 | 1429 | 99 | ||
| BT008909.1 | - | 10 - 11 | 1409 | 99 | ||
More Information
Set Membership: |
|

HAG1
Genomic Map