Assay Details
| Gene Symbol: HAG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115318.3 | NP_567002.1 | 6 - 7 | 1248 | 99 | |
| GenBank mRNA | AF338768.1 | - | 6 - 7 | 1078 | 99 | |
| BT002057.1 | - | 6 - 7 | 1081 | 99 | ||
| BT008909.1 | - | 6 - 7 | 1061 | 99 | ||
More Information
Set Membership: |
|

HAG1
Genomic Map